Would you like to log in again?
SureSTART™ Glass High Recovery Screw Top 1.7 ml Microvials for <2 ml Samples, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6ASV9-V1F 6ASV9-V1T 6ASV9-V1
TS6ASV9-V1PK 200.87 USD InStock
TS6ASV9-V1 TS6ASV9-V1T TS6ASV9-V1F
SureSTART™ Glass High Recovery Screw Top 1.7 ml Microvials for <2 ml Samples, Level 2 High-Throughput Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 1.7 ml high recovery glass screw top microvials to maximize the injection volume for <2 ml samples in high-throughput applications.
With dimensions of 12×32 mm, these microvials have an internal conical reservoir that provides a total usable volume of 1.5 ml and a residual volume of 4 µl. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty