Would you like to log in again?
SureSTART™ Crimp Caps, 11 mm, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SureSTART™
6PCCMST101X 6PCC11ST101 6PCC11TST
TS6PCCMST101XPK 83.9 USD InStock
TS6PCCMST101X TS6PCC11TST TS6PCC11ST101
SureSTART™ Crimp Caps, 11 mm, Level 3 High Performance Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 11 mm crimp caps with crimp vials that have an 11 mm opening, including SureSTART 2 ml glass crimp vials and glass crimp microvials for <2 ml samples (Level 3). Select level 3 products for ultimate performance and sensitivity from methods, while maintaining the highest levels of sample security and integrity.
Choose from aluminum and magnetic steel caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Use Thermo Scientific™ manual or electronic crimpers and decappers for easy application and removal of crimp caps.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty