Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SureSTART™ Glass Crimp Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SureSTART™
6PCV11-1P 6PCV11-1PF 6PCV11-1PT
TS6PCV11-1PEA 22.79 USD InStock
TS6PCV11-1P TS6PCV11-1PT TS6PCV11-1PF
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials, performance level 3, for high performance applications, when sensitivity is a must. Our level 3 vials are our highest performance vials, designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding GC-MS. These 2 ml clear glass crimp vials are specification certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm crimp caps and septa to seal these vials. Seal and remove your crimp caps using Thermo Scientific manual and electronic crimpers and decrimpers. These vials meet all requirements of US, EU, JPN Pharmacopeia. Buy in bulk for greater savings.

  • Performance level 2
  • SureSTART vials & cap closures for high throughput applications
  • Get the performance you need to deliver high quality data with an uninterrupted workflow
  • Our performance level 2 range has very low metal content and extractables and leachables, so that you have the assurance that each product conforms to the same high standards you are working with to meet to your regulatory requirements
  • If you need products that support routine analyzes, our performance level 2 products are here for you select the products with two waves

About Thermo Scientific SureSTART Vials, Caps, and Kits
Thermo Scientific SureSTART vials and caps are available in three performance levels. Now you can choose the products that best suit your analytical, performance, and budget needs.
Level 1 vials and caps are for everyday applications and are suitable for QA/QC applications using core detectors.
Level 2 vials, caps, and kits are for high throughput applications. These vials have very low metal content, extractables, and leachables to deliver high quality data with an uninterrupted workflow.
Level 3 vials, caps, and kits are for high performance applications. Use level 3 for your most valuable samples (and analytes prone to glass adsorption) to deliver consistent, reproducible results for your most demanding analyzes.

Certifications: Specification certified.
All Thermo Scientific SureSTART vials are dimensionally verified.
Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances.
Uniform and defect-free.
Lot traceable.
100% traceable through the manufacturing process.
Certificate of Conformance available by lot number upon request.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty