Would you like to log in again?
SureSTART™ GOLD-Grade Glass Screw and Crimp Top 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PSV9-1PGF 6PSV9-1PG 6PSV9-1PGT 6PCV11G
TS6PCV11GPK 47.27 USD InStock
TS6PCV11G TS6PSV9-1PGT TS6PSV9-1PGF TS6PSV9-1PG
SureSTART™ GOLD-Grade Glass Screw and Crimp Top 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml GOLD-grade clear glass screw and crimp top vials, performance level 3, when analyzing analytes such as polar compounds that strongly adsorb on the glass surface or are sensitive to sample pH changes.
These level 3 vials for high performance applications are made of low-adsorption GOLD-grade glass and offer; highest glass purity available; lowest possible surface activity; ultra-low adsorption and pH shift for ultra trace analysis; level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes. They are specification certified to ensure their physical dimensions and autosampler compatibility, and meet all requirements of US, EU, JPN Pharmacopeia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty