Would you like to log in again?
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PCV20-1FF 6PCV10-1F 6PCV10-1FT 6PCV6-1FF 6PCV20-1F 6PCV10-V1 6PCV10-1FF 6PCV20-1R 6PCV20-1RF 6PCV6-1FT 6PCV20-1FT 6PCV20-1RT 6PCV6-1F
TS6PCV10-1FFCS 2239.5 USD InStock
TS6PCV10-1FF TS6PCV20-1RF TS6PCV10-1FT TS6PCV20-1RT TS6PCV10-1F TS6PCV6-1FT TS6PCV20-1R TS6PCV6-1FF TS6PCV20-1F TS6PCV6-1F TS6PCV20-1FF TS6PCV20-1FT
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ glass crimp top headspace vials, performance level 3, for your volatile gas analyzes.
Available in 6, 10, and 20 ml clear glass formats, these headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyzes. This is a performance level 3 vials, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes. They are specification certified to ensure physical dimensions and autosampler compatibility. These vials meet all requirements of US, EU, JPN Pharmacopeia. Use 20 mm crimp caps and septa to seal these headspace vials. Buy in bulk for greater savings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty