Would you like to log in again?
SureSTART™ TOC Certified Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PK40TOC10 6PK40TOC20
TS6PK40TOC20PK 404.01 USD InStock
TS6PK40TOC20 TS6PK40TOC10
SureSTART™ TOC Certified Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Vials
Ensure that your environmental analyses meet government regulations with our level 3 Thermo Scientific™ SureSTART™ TOC certified screw vial and cap kits.
Select Level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Our TOC certified kits include 40 ml glass screw vials and pre-assembled polypropylene screw with bonded, high purity silicone/PTFE septa. Each lot of TOC vials is tested and certified to contribute background Total Organic Carbon levels of <10 or <20 ppb.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty