Would you like to log in again?
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PME04C1 6PME04C1F 6PME03C1SPF 6PME03C1SSPF 6PME03C1SSP 6PME03C1SPT 6PME03C1SP 6PME04C1T 6PME05F1 6PME03C1SSPT
TS6PME04C1EA 59.24 USD InStock
TS6PME04C1 TS6PME04C1F TS6PME03C1SP TS6PME04C1T TS6PME05F1 TS6PME03C1SSP TS6PME03C1SSPF TS6PME03C1SSPT TS6PME03C1SPT TS6PME03C1SPF
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Vials
Use Thermo Scientific™ SureSTART™ performance level 3 inserts with 2 ml vials when analysing <2 ml samples to maximise the injection volume. These glass inserts have a small total volume capacity of <400 μl, with a minimum residual volume of 2 µl.
When placed and sealed inside a 2 ml vial, microsamples can be used in conventional autosampler trays. Choose from polyspring conical, pulled point conical, or flat bottom insert formats. Conical inserts are most effective with residual volumes of 2 to 4 μl. These inserts are performance level 3, which means they are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are specification certified to ensure physical dimensions and meet all requirements of US, EU, JPN Pharmacopeia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty