Would you like to log in again?
SureSTART™ Precision Screw Caps, 18 mm, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PMSC18-ST2T 6PMSC18-ST2 6PMSC18-ST2F 6PMSC18-STHT 6PMSC18-STH 6PMSC18-STHF
TS6PMSC18-STHTCS 1493.03 USD InStock
TS6PMSC18-STHT TS6PMSC18-ST2 TS6PMSC18-STHF TS6PMSC18-ST2T TS6PMSC18-STH TS6PMSC18-ST2F
SureSTART™ Precision Screw Caps, 18 mm, Level 3 High Performance Applications, Thermo Scientific
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 18 mm precision screw caps with screw vials that have an 18 mm opening, including our SureSTART 10 ml and 20 ml screw top headspace vials (level 3). Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from magnetic steel caps with all-purpose silicone/PTFE septa designed to reduce autosampler needle issues. Select high temperature septa to allow for heating to over 250 °C without excessive septum softening or the release of volatile vapors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty