Would you like to log in again?
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
SureSTART™
6PSC9STBS1F 6PSC9STB1 6PSC9STB1F 6PSC9ST1RT 6PSC9TSTRF 6PSC9TSTRT 6PSC9TST 6PSC9STS1F 6PSC9TSTF 6PSC9TSTT 6PSC9ST1RF 6PSC9ST101X 6PSC9STS1 6PSC9ST1 6PSC9ST1F 6PSC9STS1R 6PMSC9ST1 6PSC9STS1RT 6PSC9STBS1 6PSC9TSTR 6PSC9ST101 6PSC9STBS1T 6PSC9STS1RF 6PSC9STS1T 6PSC9STB1T 6PSC9ST1R 6PSC9ST1T
TS6PSC9TSTRPK 51.99 USD InStock
TS6PSC9TSTR TS6PSC9TSTT TS6PSC9STS1RT TS6PSC9STB1T TS6PSC9STS1F TS6PSC9TSTF TS6PSC9ST101 TS6PSC9STS1RF TS6PSC9ST1RT TS6PSC9STB1F TS6PSC9TSTRT TS6PSC9STBS1F TS6PSC9STBS1 TS6PSC9ST1RF TS6PSC9STS1 TS6PSC9TSTRF TS6PSC9ST1 TS6PSC9STB1 TS6PSC9TST TS6PSC9ST1F TS6PMSC9ST1 TS6PSC9STBS1T TS6PSC9STS1R TS6PSC9STS1T TS6PSC9ST101X TS6PSC9ST1R TS6PSC9ST1T
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Closures Chromatography Vial Closures
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including SureSTART™ 2 ml glass screw vials, 2 ml gold-grade glass screw vials, total recovery glass screw microvials for <2 ml samples, 1.7 ml high recovery glass screw vials, and glass screw microvials for <2 ml samples (Level 3). Select Level 3 products for ultimate performance and sensitivity from methods, while maintaining the highest levels of sample security and integrity.
Choose from polypropylene caps with all-purpose silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. AVCS™ caps incorporate advanced vial closure system technology to ensure optimal seal compression when closing a vial. Use AVCS™ screw caps with SureStop™ vials for effective seals, consistent data, and increased sample throughput.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty