Would you like to log in again?
SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™
6PSV9-TR1T 6PSV9-TR1F 6PSV9-CHV1 6PSV9-SV1 6PSV9-TR1
TS6PSV9-TR1PK 283.6 USD InStock
TS6PSV9-TR1 TS6PSV9-CHV1 TS6PSV9-SV1 TS6PSV9-TR1F TS6PSV9-TR1T
SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Vials
Choose Thermo Scientific™ SureSTART™ 1.5 ml total recovery glass screw top microvials, performance Level 3, when you need to maximize the injection volume for <2 ml samples.
Available in clear and amber glass, this one-piece total recovery microvials has a very low residual volume due to a specially-shaped deep and narrow inner reservoir which collects the liquid in the base so that compatible autosamplers can collect all but 1 to2 µl of sample while having a total sample volume of 1.2 ml and a residual volume of 1 µl. The outer dimensions for these vials are 12×32 mm. These performance Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty