Would you like to log in again?
DNAPac™ RP Column, Thermo Scientific
Supplier: Thermo Scientific
DNAPac™
088921 088923 088925 088919 088920 950-00 088924 303324
10841-614EA 1075.4 USD InStock
10841-614 10841-612 TS950-00 10841-616 10841-562 76420-872 10841-566 10841-564
DNAPac™ RP Column, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ DNAPac™ RP is a reversed-phase (RP) liquid chromatography column designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC/UV or LC/MS.
  • For ion-pair reversed-phase (IP-RP) separations
  • High resolution and high throughput
  • Excellent MS compatibility
  • Wide operating pH range (0 to 14)
  • High temperature stability (up to 110 ºC)
DNAPac™ RP are designed for ion-pair reversed-phase (IP-RP) separations of both oligonucleotides and double-stranded nucleic acids. The unique column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10 k base pairs.
The DNAPac RP column is based on spherical wide-pore 4 μm polymer resin. The phase is highly stable to extremes of pH (0 to 14), and temperature (5 to 110 °C), and operates with mobile phases compatible with direct mass spectrometry. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments. DNAPac RP reversed phase (RP) column is specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents.
Ordering information: Guard cartridges require use of the corresponding guard holder.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty