Would you like to log in again?
SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SureSTART™
CHCC11-10 CHCC11-10F CHCC11-30 CHCC11-10T CHCC11-30F CHCC11-30T
TSCHCC11-10TCS 123.48 USD InStock
TSCHCC11-10T TSCHCC11-10F TSCHCC11-30T 76307-606 TSCHCC11-30F 76307-604
SureSTART™ Caps for 2 ml Glass Crimp Top Vials, Level 1 Everyday Analysis, Thermo Scientific
Closures Chromatography Vial Closures
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types.
These 2 ml glass crimp top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN pharmacopia. Seal these crimp vials with 11 mm aluminum crimp caps and septa using a manual or electronic crimper tool.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty