Would you like to log in again?
TechniSat® Gown Room Wiper, Texwipe®
Supplier: Texwipe
TechniSat®
TX1048
TW-TX1048CS 410.82 USD InStock
TW-TX1048
TechniSat® Gown Room Wiper, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Pre-wetted cellulose and polyester blend wiper for effective, economical wipedown of articles brought into the cleanroom.
Pre-wetted with the optimal amount of 0.2 µm-filtered, blended 6% semiconductor-grade isopropyl alcohol (IPA) and 94% deionized water solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty