Would you like to log in again?
Dry Cotton Cleanroom Wipers, Texwipe®
Supplier: Texwipe
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TX312 TX306 TX304 TX309
TW-TX312CS 1430.74 USD InStock
TW-TX312 TW-TX306 TW-TX304 TW-TX309
Dry Cotton Cleanroom Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipers are made from 100% cotton material with a cut edge, cleanroom manufactured.
  • Designed for use on abrasive surfaces
  • Will not easily snag or abrade releasing particles and fibers into the process or environment
  • Bias-cut to minimize edge shedding and fraying
  • Made from sustainable and renewable materials grown and processed in the US
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Autoclave safe
The wipers are constructed from long staple 100% cotton yarn to reduce free floating fibers on the surface while still providing excellent wiping and cleaning. Minimizes cleanroom contamination with low-lint surface and edges protected by our ULP (Ultra-Low Particulate) treatment.
Cleanroom Environment: ISO Class 6 - 8, Class 1,000 - 100,000, EU Grade B - D
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty