Would you like to log in again?
AlphaMop™ Polyester Mop, Texwipe®
Supplier: Texwipe
AlphaMop™
TX7118 TX7123 TX7026 TX7118F STX7118F TX7118M TX7108H TX7108 TX7108AH STX7118M
TW-TX7026CS 912.36 USD InStock
TW-TX7026 TWTX7118F TWTX7108 TWSTX7118F 10127-118 TW-TX7118 10127-120 10127-132 10127-154 TWT7118M
AlphaMop™ Polyester Mop, Texwipe®
Mops
Ideal for cleaning walls, floors, and other flat surfaces in critical environments
AlphaMop™ covers TWTX7026 are pre-wetted with 6% isopropyl alcohol. The case purchase includes two foam pads. Replacement pack TW-TX7118 contains 150 polyester heads and 6 foam pads.
The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fiberglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. Lightweight mop maintains ultralow generation of particles and extractables. Bag-Within-A-Bag® packaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty