Would you like to log in again?
SWAB COTTON POLYSTYRENE HANDLE 500/BG
Supplier: Texwipe
Swab^ Swab, 100% USP-grade cotton head with polystyrene handle, Dry Collection and Transport System, length : 155 mm, Non Sterile, Swabs are bonded securely to handle by an aqueous-based adhesive, Handle Color: White, 1 swab/tube
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SWAB COTTON POLYSTYRENE HANDLE 500/BG TW-TX705P TX705P Bag of 500 Retrieving
Case of 10000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty