Would you like to log in again?
AlphaWipe® Cleanroom Wipers
Supplier: Texwipe
AlphaWipe®
TX1025 TX1013 211172 TX1009B TX1009 TX1004
TWTX1009CS 904.27 USD InStock
TWTX1009 82020-900 TWTX1004 TWTX1025 76244-880 TWTX1013
AlphaWipe® Cleanroom Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
AlphaWipe® is an ultraclean and highly adsorbent wiper ideal for critical environments.
  • Soft and nonabrasive
  • Excellent for sensitive surfaces
Made from 100% continuous-filament polyester, AlphaWipe® wipers are constructed in a double-knit, no-run, interlock pattern.
Ordering information: Wipers are cleanroom laundered and packed in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty