Would you like to log in again?
TechniSat® Presaturated Wipers, Texwipe®
Supplier: Texwipe
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TechniSat®
TX1041 TX1065 TX1045 TX1067 TX1041 TX1045 TX1065 TX8488
TWTX1045CS 611.7 USD InStock
TWTX1045 TWTX1067 TW-TX1041 CATW-TX1041 TW-TX1065 TWTX8488 CATWTX1045 CATWTX1065
TechniSat® Presaturated Wipers, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These convenient wipers provide portion control and reduced potential of spillage in and out of the cleanroom.
  • Fibers are blended by a hydroentanglement procedure
  • Uses water instead of chemicals to bind
  • Excellent bidirectional strength and a very low extractable level
  • Wipers are controlled for traceability
The blended material of 55% cellulose and 45% polyester provides the high absorbency of a natural fiber with the cleanliness and strength of a synthetic material. For non sterile wipers, the 70% semiconductor-grade isopropyl alcohol is filtered to 0.2 µm and diluted to 70% concentration with 18 megohm-cm deionized water. Nonsterile wipers are available in FlexPacks or recloseable bags. Sterile wipers are saturated with USP grade isopropyl alcohol and packaged in flexpacks. The flexpacks are then double-bagged to maintain sterility.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty