Would you like to log in again?
Vectra® QuanTex® Wiper, Texwipe®
Supplier: Texwipe
Vectra® QuanTex®
TX1080
TWTX1080BG 107.79 USD InStock
TWTX1080
Vectra® QuanTex® Wiper, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Double-knit 100% continuous-filament polyester wiper offers high sorbency performance.
  • All four edges are thermally sealed, forming a border to prevent particle and fiber release
  • Steam autoclavable for use in sterile environments
  • Wiper resists abrasion under rigorous use or when wiping rough surfaces
Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Certifications: Features bag-to-bag lot traceability, from wiper to base fiber.
Ordering information: To reduce particulation, wiper is Vectra processed with online statistical process control, machine vision inspection, and robotic stacking. Packed in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty