Would you like to log in again?
TechniCloth® II Wipers, Texwipe®
Supplier: Texwipe
TechniCloth®
TX1118 TX1109 TX1109 TX1112
TWTX1109CS 558.16 USD InStock
TWTX1109 TWTX1118 76244-858 TWTX1112
TechniCloth® II Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for critical environments where low particulate levels and high absorbency are required.
  • Provides the high absorbency of a natural fiber with the strength of a synthetic fiber
  • Finish is non-ionic and doesn’t extract into commonly used solvents
Wipers are constructed of a clean, strong, hydroentangled blend of 45% polyester and 55% cellulose. Wipers are also treated with an ultra-low particle finish that gives the wipers significantly lower particle generation. Suitable for use in ISO Class 6 (FED-STD-209E Class 1000/M4.5) environments.
Ordering information: Cut and packed in an ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom and packaged in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty