Would you like to log in again?
BetaWipe™ Wiper, Texwipe®
Supplier: Texwipe
BetaWipe™
TX2009
TWTX2009CS 619.58 USD InStock
TWTX2009
BetaWipe™ Wiper, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wiper is made from thermally bonded polypropylene and cellulose.
  • Soft, pillowy texture makes it ideal for wiping abrasion-sensitive surfaces
  • Surface texture allows for greater sorption of liquids - wiper can absorb five times its weight in water
Cellulose contributes to wiper's high sorption while the polypropylene provides resistance to strong acids. Suitable for use in ISO Class 6 (FED-STD-209E Class 1000/M4.5) environments. Packed in an ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom.
Ordering information: Packaged in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty