Would you like to log in again?
AlphaSorb® HC Wipers
Supplier: Texwipe
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaSorb®
TX2412 TX2418 TX2409
TW-TX2409BG 46.08 USD InStock
TW-TX2409 TWTX2418 TWTX2412
AlphaSorb® HC Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
AlphaSorb® HC (high capacity) two-ply, knitted, 100% polyester wipers are engineered to combine high sorbency with low particle and extractable levels.
  • Autoclavable
The large surface area is especially suited for use in the pharmaceutical and biotechnology industries where large, highly sorptive wipers are required for applying and removing cleaning, sterilizing, and disinfecting solutions.
Certifications: Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Ordering information: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom and packed in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty