Would you like to log in again?
SterileWipe™ High Sorbency II Wiper, Texwipe®
Supplier: Texwipe
SterileWipe™
TX3210
TWTX3210CS 539.38 USD InStock
TWTX3210
SterileWipe™ High Sorbency II Wiper, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Absorbent cellulose/polyester wiper is designed for sterile environments.
Chemically resistant to withstand exposure to common cleaning and disinfecting solutions such as bleaches, phenols, and quaternary ammonium compounds. Packaging permits alcohol wipe down of exterior bag before introduction of the inner bag into sterile suites.
SterileWipe™ High Sorbency II is a nonwoven 55% cellulose/45% polyester blend wiper offering both durability and absorbency. SterileWipe™ High Sorbency II is an excellent general-purpose sterile wiper in areas where economy and cleanliness are of critical importance. It offers an efficient balance of low particle counts, low extractables, adsorbency, and good solvent compatibility.
Ordering information: Packaged 20 wipers per inner bag, 5 inner bags per outer bag, 5 outer bags per case.
Packaging: Each package is gamma irradiated to 10⁻⁶ Sterility Assurance Level.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty