Would you like to log in again?
SterileWipe™ Low Particle 10 Wiper, Texwipe®
Supplier: Texwipe
SterileWipe™
TX3212
TWTX3212CS 1110.74 USD InStock
TWTX3212
SterileWipe™ Low Particle 10 Wiper, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
SterileWipe™ Low Particle 10 Wiper is a 100% continuous-filament polyester wiper that is low in particle generation and extractable levels and provides excellent cleaning performance in critical sterile environments
Ideal for use when bacteriological contamination, particulates, and extractables are critical factors, and ideal for environments where low particulate levels are most critical. Suitable for use in ISO Class 1-3 environments.
Each package is gamma irradiated to 10–6 Sterility Assurance Level.
Designed for cleaning sterile environments and dispensing solutions.
Ordering information: Packaged 10 wipers per pack, 10 packs per bag in Solvent Safe packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty