Would you like to log in again?
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
Supplier: Texwipe
TX3252 TX3252
TWTX3252CS 1006.62 USD InStock
CATWTX3252 TWTX3252
STERILE ALPHASAT® featuring AlphaSorb HC®, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Sterile AlphaSat® incorporates Texwipe's highly sorptive AlphaSorb® HC polyester wipers plus 70% IPA.
  • Constructed from a unique two-ply, pinsonic fabric
  • 100% knitted continous-filament polyester
Chosen for their low particle-generating characteristics and low extractable levels, these wipers offer superior strength in a continous-filament knitted substrate.
This wiper is saturated with a 0.2 μm filtered, 70% isopropyl alcohol/30% deionized water solution. Packaging permits alcohol wipedown of exterior bag before introduction of the inner bag into sterile suites. Gamma irradiated to 10⁻⁶ Sterility Assurance Level. Fibers (>100 μm) are 230 fibers/m². The isopropyl alcohol extractant is 0.09 g/m². The deionized water extractant is 0.05 g/m².
Sterile AlphaSat® with AlphaSorb® HC is ideal for alcohol wipedowns in the most critical areas of pharmaceutical sterile environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty