Would you like to log in again?
Vertex™ High Sorption Cleanroom Wipers
Supplier: Texwipe
Vertex™
TX42 TX49P TX42 TX49
TWTX49CS 994.9 USD InStock
TWTX49 TWTX49P TWTX42
Vertex™ High Sorption Cleanroom Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These 100% continuous-filament polyester, sealed-edge wipers are processed in a hands-free micro-environment on the Vertex™ manufacturing system.
  • Vertex wipers are available pre-wetted in 70% USP-grade isopropyl alcohol
  • Packaged in a standup reclosable slider bag for ease of use and a small footprint
The wiper's unique design provides a very soft feel and high sorptive capacity in a single ply wiper. The wipers are manufactured in an ISO Class 3 environment with a small physical footprint which provides for a more sustainable process. Made in the USA, Vertex wipers are tested quarterly for cytotoxicity and endotoxins. Having the same wiper and manufacturing platform allows for single product qualification to represent all High Sorption Series products saving time and expense.
Ordering information: Vertex wipers are also available in the high durability series and the microdenier series.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty