Would you like to log in again?
TechniCloth® Cleanroom Wipers
Supplier: Texwipe
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TechniCloth®
TX606 TX609 TX612 TX604
TWTX609CS 265.4 USD InStock
TWTX609 TWTX612 TWTX604 TWTX606
TechniCloth® Cleanroom Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for cleaning computer and electronic equipment.
  • Will not scratch or abrade delicate precision components
  • Non-woven cleanroom wipers
  • Combine the soft strength of polyester with the high absorbency of cellulose
Excellent bidirectional strength and extremely low levels of solvent extractables. Suitable for use in ISO Class 6 (FED-STD-209E Class 1000/M4.5) environments.
Ordering information: Cut and packed in an ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom and packaged in SolventSafe Bag-Within-A-Bag® packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty