Would you like to log in again?
CleanTips® Cotton Foam-Covered Swab, Texwipe®
Supplier: Texwipe
CleanTips®
TX700B
TWTX700BCS 855.84 USD InStock
TWTX700B
CleanTips® Cotton Foam-Covered Swab, Texwipe®
Swabs Swabs for Cleaning
The Foam-Covered Cotton Bud is a highly sorbent, general purpose swab for cleaning smaller surfaces and hard-to-reach areas
Its open-cell polyurethane foam head possesses excellent absorptive capacity and cushioning. This swab has the additional advantage of being seamless, with no exposed edges. Ideal for cleaning with solvents such as IPA, applying lubricants and other liquids, and removing excess materials. The foam surrounds an internal cotton bud core, which provides cushioned support and additional sorbency. The long hardwood handle allows the head to access recessed areas. Low fiber and particle generation. Good chemical resistance.
Ordering information: Available as 500/bag (10 inner bags of 50) or 2500/case quantities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty