Would you like to log in again?
FoamWipe™ Wiper, Texwipe®
Supplier: Texwipe
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FoamWipe™
TX704
TWTX704CS 525.86 USD InStock
TWTX704
FoamWipe™ Wiper, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
An excellent choice for critical environments where the characteristics of foam are desired and low particulation is essential.
The fully reticulated, 100 pore-per-inch polyurethane foam is cut with a precision hot-wire die to ensure dimensional uniformity. The hot-wire die uses no cutting oils and provides a razor-clean, noncontaminating cut.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty