Would you like to log in again?
CleanTips® Keyboard Foam Swab, Texwipe®
Supplier: Texwipe
CleanTips®
TX706A
TWTX706ACS 615.55 USD InStock
TWTX706A
CleanTips® Keyboard Foam Swab, Texwipe®
Swabs Swabs for Cleaning
The keyboard swab has been specifically designed to clean keyboards and keypads
Swab features elongated, flexible head on rounded, easy-to-hold polypropylene handle. Its 100ppi closed-cell polyurethane foam head possesses excellent durability for scrubbing stubborn dirt. A thin internal paddle provides support when cleaning between keys. Thermally-bonded. Head will not crack or chip.
The thick, light green polypropylene handle enables good control and ease of use.
Can be used in a wide variety of industrial applications, from general assembly to cleanrooms. Suitable for applications requiring high absorbency, low particle generation, and uniform dispersing of solvents. The foam's inherent softness and low abrasion protect delicate components.
Ordering information: Packaged as a bag of 100 (two inner bags of 50 swabs) or 1000 per case quantities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty