Would you like to log in again?
Vectra® QuanSat® LT TX8691 Pre-Wetted Cleanroom Wipers, Non-Sterile, Texwipe®
Supplier: Texwipe
Vectra® QuanSat®
TX8691
TWTX8691CS 282.26 USD InStock
TWTX8691
Vectra® QuanSat® LT TX8691 Pre-Wetted Cleanroom Wipers, Non-Sterile, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Vectra® Alpha® 10 LT pre-wetted cleanroom wiper with 6% IPA is made from low titanium dioxide (TiO2) 100% polyester, processed on Texwipe’s automated cleaning system.
  • Reclosable bag with internal security seal
  • Designed for use on abrasive surfaces.
  • Excellent chemical resistance for compatibility with a varierty of solutions
  • Vectra® sealed border technology provides low levels of ions, NVRs (non-volatile residues), particles and fibers
The Vectra® Alpha® 10 LT is designed with a low level of titanium dioxide (TIO2), safe for processes that are affected by the presence of TIO2. Its sealed border technology allows for a more safe and thorough process in wiping and cleaning surfaces, equipment and parts. Furthermore, The Vectra® Alpha® 10 LT is suitable in applying and removing lubricants, adhesives, residues, and other solutions including disinfectants.
Cleanroom Environment: ISO Class 1 - 6, Class <1 - 1,000, EU Grade A - D
Packaging: Packaged in an easy-to-use flatpack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty