Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PolySat® 70% IPA Presaturated Wipers, Texwipe®

Supplier: Texwipe

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PolySat®
TX8723 TX3216 TX1040 TX3216 TX8727 TX1051 TX1051 TX3213
TWTX8727CS 362.96 USD InStock
TWTX8727 TWTX1040 TWTX1051 TW-TX3216 TWTX8723 CATW-TX3216 CATWTX1051 TWTX3213
PolySat® 70% IPA Presaturated Wipers, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
POLYSAT® wipers are made from 100% polypropylene and pre-wetted with 70% IPA.

Nonsterile wipers are saturated with 70% semiconductor-grade isopropyl alcohol that is filtered to 0.2 µm and diluted with 18 mega ohm deionized water. Sterile wipers are saturated with USP-grade isopropyl alcohol, packaged in flexpacks, and double-bagged to maintain sterility. Gamma irradiated to 10⁻⁶ Sterility Assurance Level.

Wiper TWTX8727 was engineered for small spaces and easy storage, folded and encased in a locking container. This product features a folded wiper (6×4" and opens to 6×11") in a secure, locking container for reduced evaporation and easily refill.

The container for wiper TWTX8727 is engineered for easy, one-hand operation (from package to application) and the wipe offers low particle generation and low levels of solvent extractables.

These soft wipers lend themselves to general purpose cleaning on sensitive surfaces.

Ordering information: Wiper TWTX8727 is packaged as 75 wipers/refill bag and contains 20 refill bags plus one container/case.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty