Would you like to log in again?
ULTRAgrade®, Single Element Standards for ICP, Beryllium, Agilent Technologies
Supplier: Agilent Technologies
ICP-104 ICP-004
UL-ICP-004EA 77.9 USD InStock
UL-ICP-004 UL-ICP-104
ULTRAgrade®, Single Element Standards for ICP, Beryllium, Agilent Technologies
Standards Cationic Standards Beryllium Standards
The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system. It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).
It is traceable to NIST standard reference materials (SRM) whenever possible.
ULTRAgrade® documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty