Would you like to log in again?
Magnesium, single element standards for ICP, Ultra Scientific
Supplier: Agilent Technologies
ICP-012 ICP-112
UL-ICP-012EA 69.07 USD InStock
UL-ICP-012 UL-ICP-112
Magnesium, single element standards for ICP, Ultra Scientific
Standards Cationic Standards Magnesium Standards
The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system. It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).
It is traceable to NIST standard reference materials (SRM) whenever possible.
ULTRAgrade® documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty