Would you like to log in again?
Phosphorus Single Element Standards for ICP and ICP-MS, Ultra Scientific
Supplier: Agilent Technologies
ICP-015 ICP-115
UL-ICP-015EA 73.59 USD InStock
UL-ICP-015 UL-ICP-115
Phosphorus Single Element Standards for ICP and ICP-MS, Ultra Scientific
Standards Cationic Standards Phosphorus Standards
The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system. It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).
It is traceable to NIST standard reference materials (SRM) whenever possible.
Documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty