Would you like to log in again?
ULTRAgrade® Lead, Single Element Standards for ICP, Ultra Scientific, Agilent Technologies
Supplier: Agilent Technologies
ULTRAgrade®
ICP-082 ICP-182
UL-ICP-082EA 67.49 USD InStock
UL-ICP-082 UL-ICP-182
ULTRAgrade® Lead, Single Element Standards for ICP, Ultra Scientific, Agilent Technologies
Standards Cationic Standards Lead Standards
The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system
It is traceable to NIST standard reference materials (SRM) whenever possible.
ULTRAgrade® documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.
It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty