Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EPA Method 527, Ultra Scientific
Supplier: Agilent Technologies
PPM-527B-1 ISM-710-1 PPM-527A-1 PPM-527C-1
ULPPM-527C-1EA 930.58 USD InStock
ULPPM-527C-1 ULISM-710-1 ULPPM-527B-1 ULPPM-527A-1
EPA Method 527, Ultra Scientific
Standards Environmental Analysis Standards
EPA Method 527 is used to determine SOCs
The surrogate standard solution contains three analytes at 500µg/mL in actetone. Pesticide mixture #1 and #2 each contain five analytes at 500µg/mL in ethyl acetate. PDBE mixture contains 16 analytes at 500µg/mL in ethyl acetate. Volume: 1mL.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty