Would you like to log in again?
Vertical Rods, WaterSaver Faucet
Supplier: WaterSaver Faucet
AAP110-018 AAP110-024 AAP110-036
VF-0549-24EA 52.61 USD InStock
VF-0549-24 VF-0549-36 VF-0549-18
Vertical Rods, WaterSaver Faucet
Support Rods and Frames
Vertical rods are machined with the top end rounded and the bottom end tapered to fit in to a rod socket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty