Would you like to log in again?
N Series Fume Extractor Fans and Manual Starters, Nederman
Supplier: Nederman
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
14502437 14520529 14510929 14502337 14510223 14510229 14502637 14511022 14502237 14502137 14502537 14510321 14503137 14510222 14503037 14511021 14522229
46102-232EA 4199.81 USD InStock
46102-232 46102-242 46102-220 46102-222 46102-244 46102-250 46102-230 46102-240 46102-228 46102-236 46102-214 46102-224 46102-246 46102-238 46102-216 46102-248 VF-510222
N Series Fume Extractor Fans and Manual Starters, Nederman
Fume Extractors
Designed for Nederman extractor arms
The simple and powerful fans extract noxious fumes and vapors for various applications. Single or three-phase 60Hz fans are available. The fan starters feature a lockable on/off switch and can be used for both single and three-phase fans. Fans and fan starters are also equipped with motor overload protectors.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty