Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories for VWR® Modular Workstations for 72×30×30" and 72×30×36"

Supplier: Avantor
ALSFP72 ALS12P72 ALSFL72 AP72
VF-ALSFL72EA 532.44 USD InStock
VF-ALSFL72 VF-AP72 VF-ALS12P72 VF-ALSFP72
Accessories for VWR® Modular Workstations for 72×30×30" and 72×30×36"
Furniture Tables and Workbenches

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty