Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Modular Workstations, W×D×H 72×30×36"

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

B723036E B723036F B723036EB B723036P B723036PB B723036L
VF-B723036LEA 1920.81 USD InStock
VF-B723036L VF-B723036EB VF-B723036E VF-B723036F VF-B723036P VF-B723036PB
VWR® Modular Workstations, W×D×H 72×30×36"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.

  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments

Standard work benches handle 1000lbs

The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5” x 1.5” steel frame. Shipped fully assembled. Also available in 48" and 60" widths.

of evenly distributed weight but can be configured to handle loads of 5000lbs. or more. Tables can be custom configured with numerous accessories including, 8" adjustable leg kits (for ADA compliance), locking casters, heavy-duty 2x2 steel frame, upper and lower shelves with pencil lips, backsplashes, lighting, bin rails, power strips, ESD tops, and steel drawers.

Ordering information: Select WorkBench size followed by type of work surface (epoxy, plastic laminate, ESD, phenolic). Next add appropriate accessories to the bench to complete your configuration. If you need something not on the price list, then please contact your VWR sales representative. Exact frame dimensions depend upon reveal and thickness of work surface to be added.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty