Would you like to log in again?
Stainless Steel Drip Troughs, Interdyne Inc
Supplier: Interdyne
HDT-42-2 HDT-18-2 HDT-36-2 HDT-24-2 HDT-32-2 HDT-60-2 HDT-20-2 HDT-30-2 HDT-48-2
VF-HDT-32-2EA 162.38 USD InStock
VF-HDT-32-2 VF-HDT-20-2 VF-HDT-30-2 VF-HDT-42-2 VF-HDT-60-2 VF-HDT-18-2 VF-HDT-36-2 VF-HDT-24-2 VF-HDT-48-2
Stainless Steel Drip Troughs, Interdyne Inc
Trays/Dishes/Pots Spillage Trays/Pans
All stainless steel construction, made from 20 gauge material, Type 304 stainless steel with No. 4 finish.
  • 1" mounting flange.
  • 3 ft flexible drain hose included.
4 finish.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty