Would you like to log in again?
SAFE-T-ZONE® Emergency Stations, Speakman®
Supplier: Speakman
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SAFE-T-ZONE®
SE-693 SE-695 SE-690 SE-697
VF-SE-690EA 2904.91 USD InStock
VF-SE-690 VF-SE-693 VF-SE-697 VF-SE-695
SAFE-T-ZONE® Emergency Stations, Speakman®
First Aid Supplies Showers
Emergency stations combine a deluge shower with a eyewash or eye/face wash unit.
  • Eyewashes feature dual aerated spray heads with stay-open ball valve
  • Eye/face washes feature dual spray heads with automatic flow control
All eyewashes and eye/face washes activate via a stainless steel push handle and include flip-top dust covers. Choose from plastic bowl or stainless steel bowl.
All feature an impeller action deluge showerhead with a high-visibility yellow pull rod, situated on a painted galvanized steel stanchion.
Certifications: Meet ANSI Z358.1 standard.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty