Would you like to log in again?
Eyesaver Drench Hose and Eyewash Combination, Model SE-920, Speakman®
Supplier: Speakman
Eyesaver®
SE-920
VF-SE-920EA 748.25 USD InStock
VF-SE-920
Eyesaver Drench Hose and Eyewash Combination, Model SE-920, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Speakman® Drench hose and eyewash combinations provide great flexibility in laboratories and other industrial areas for flushing the eyes and body.
  • Hose and eyewash combination
  • Laboratory flexibility
  • For direct flushing on contaminated areas
All units come standard with a flexible hose. Model SE-920 is a wall mounted drench hose featuring one aerated spray outlet and a 6' hose. The unit also features a self-closing valve and hook for the drench hose.
Certifications: Meets ANSI 358.1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty