Would you like to log in again?
VWR® Modular Workstations with Upper Support, W×D×H 48×30×30"
Supplier: VWR
U483030PB U483030EB
VF-U483030EBEA 4332.22 USD InStock
VF-U483030EB VF-U483030PB
VWR® Modular Workstations with Upper Support, W×D×H 48×30×30"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped with assembled frame
  • Overhead storage capability with addition of uprights
Standard work benches handle 1000lbs
The addition of two uprights (unistruts) to the VWR® Basic Work Bench provides overhead storage capability. The upper support bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame, as well as a single 12" deep adjustable overhead laminate shelf attached to two unistruts. Shipped with assembled frame; upper accessories must be attached upon arrival. Also available in 60" and 72" widths.
of evenly distributed weight but can be configured to handle loads of 5000lbs. or more. Tables can be custom configured with numerous accessories including, 8" adjustable leg kits (for ADA compliance), locking casters, heavy-duty 2x2 steel frame, upper and lower shelves with pencil lips, backsplashes, lighting, bin rails, power strips, ESD tops, and steel drawers.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty