Would you like to log in again?
Balance Table, Waller Brothers
Supplier: Waller
STD.BAL.TBL
VF7-6590-00EA 14347.67 USD InStock
VF7-6590-00
Balance Table, Waller Brothers
Furniture Tables and Workbenches
This rock-steady weighing surface is ideal for delicate microbalance procedures.
  • Balance table is made of heavy 7.6 cm (3") thick black KEMROCK® slabs
  • Connected by a steel pipe with flanges
The weight of the table minimizes floor vibrations that would be transferred to an ordinary tabletop.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty