Would you like to log in again?
Mod-Rack™ Pegboard, Interdyne
Supplier: Interdyne
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Mod-Rack™
V1824 V3030 V2424
VF9-V1824EA 754.04 USD InStock
VF9-V1824 VF9-V2424 VF9-V3030
Mod-Rack™ Pegboard, Interdyne
Racks Draining Racks
Stainless steel pegboard features interchangeable white polypropylene pegs, integral drip trough, and clear plastic drain tube
Completely autoclavable and stain-resistant. Wall bracket included.
Ordering information: Accessories including a funnel rack, drain basket, screen insert, and pipet rack are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty