Would you like to log in again?
Wall Mounted Needle Valve, WaterSaver Faucet
Supplier: WaterSaver Faucet
CT2880258WSA CT2880141WSA
VFCT2880141WSAEA 268.04 USD InStock
VFCT2880141WSA VFCT2880258WSA
Wall Mounted Needle Valve, WaterSaver Faucet
Faucets and Sinks Faucets
White, epoxy-coated, single straight pattern laboratory needle valve.
Brass construction with stainless steel needle and seat. Needle and seat are removable/replaceable for infinite service life. Black, nylon, hooded handle with color-coded index button. Mounting shank included. Rated for use at pressures up to 10.3 bar (150 psi).
Ordering information: Please specify service when ordering.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty