Would you like to log in again?
Wall-Mount Eye/Face Washes, WaterSaver Faucet
Supplier: WaterSaver Faucet
FE724 CTFEBF724
59380-016EA 826.61 USD InStock
59380-016 VFCTFEBF724
Wall-Mount Eye/Face Washes, WaterSaver Faucet
First Aid Supplies Eyewash Stations and Eyewash Bottles
Featuring four GS-Plus “Gentle Spray” heads, these eye/face washes provide a soft, wide stream of water that gently and quickly rinses contaminants away.
  • Heavy-duty pipe fittings resist corrosion
The integral, self-adjusting flow controls, located in the head, ensure even distribution of water. Units come complete with a 29.2 cm (11") stainless steel bowl and a 1.3 cm (¹/₂") brass stay-open ball valve.
Certifications: Meet ANSI Z358.1 standard.
Delivery information: Supplied with ANSI-compliant safety green emergency eye wash sign.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty