Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safety Shower Stations, WaterSaver Faucet

Supplier: WaterSaver Faucet

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SSC902 SS902 SSCBF909 SSC909 CTSSBF919 CTSS912 CTSS919
56611-008EA 1511.39 USD InStock
56611-008 VFCTSS919 VFCTSSBF919 59380-036 VFCTSS912 59380-038 59380-034
Safety Shower Stations, WaterSaver Faucet
First Aid Supplies Showers
Designed to provide complete personal protection, these corrosion-resistant safety stations feature a single water inlet that supplies both the shower and eye wash.

  • Supplied with rigid stainless steel pull rods
  • Self-regulating flow controls are positioned inside each spray head
  • Stations are provided with an ANSI-compliant sign
  • 2 versions available: Orange ABS plastic shower head and galvanized steel pipes and fittings with orange PE covers or cast brass showerhead and white epoxy/polyester hybrid powder coated brass pipe and fittings

The high volume deluge shower head has a minimum 75.7 L/min. (20 gpm) flow and 50.8 cm (20") spray pattern.

Certifications: Meets ANSI Z358.1 standard.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty